Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRSS36
Entrez Gene ID
101106230
Gene Name
serine protease 36
Gene Aliases
-
Chromosome Location
Chr.24: 27996442 - 28011754 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027961538.1 XP_027817339.1 20 - 21 3590 67

View Full Product Details