Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRO
Entrez Gene ID
106990669
Gene Name
trophinin
Gene Aliases
-
Chromosome Location
Chr.X: 53288690 - 53301517 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027963423.1 XP_027819224.1 5 - 6 1467 77

View Full Product Details