Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DNASE1L2
Entrez Gene ID
101115336
Gene Name
deoxyribonuclease 1 like 2
Gene Aliases
-
Chromosome Location
Chr.24: 1714733 - 1718496 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027962029.1 XP_027817830.1 5 - 6 1914 74

View Full Product Details