Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WNT16
Entrez Gene ID
101103229
Gene Name
Wnt family member 16
Gene Aliases
-
Chromosome Location
Chr.4: 93856232 - 93868706 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004008012.4 XP_004008061.2 1 - 2 515 61

View Full Product Details