Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FLYWCH1
Entrez Gene ID
105604619
Gene Name
FLYWCH-type zinc finger 1
Gene Aliases
-
Chromosome Location
Chr.24: 2278728 - 2301958 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027961955.1 XP_027817756.1 3 - 4 710 109

View Full Product Details