Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
USP50
Entrez Gene ID
101106690
Gene Name
ubiquitin specific peptidase 50
Gene Aliases
-
Chromosome Location
Chr.7: 61573901 - 61591007 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027972250.1 XP_027828051.1 5 - 6 907 113

View Full Product Details