Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
SRM
Entrez Gene ID
101112338
Gene Name
spermidine synthase
Gene Aliases
-
Chromosome Location
Chr.12: 45970148 - 45974215 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027975643.1 XP_027831444.1 7 - 8 1025 104

View Full Product Details