Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARRDC5
Entrez Gene ID
101110504
Gene Name
arrestin domain containing 5
Gene Aliases
-
Chromosome Location
Chr.5: 17741862 - 17760532 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015095879.2 XP_014951365.1 2 - 3 563 97

View Full Product Details