Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
XRCC2
Entrez Gene ID
101105407
Gene Name
X-ray repair cross complementing 2
Gene Aliases
-
Chromosome Location
Chr.4: 125089078 - 125113309 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004008182.4 XP_004008231.1 2 - 3 211 105

View Full Product Details