Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACTC1
Entrez Gene ID
101111554
Gene Name
actin alpha cardiac muscle 1
Gene Aliases
-
Chromosome Location
Chr.7: 29995536 - 30000833 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004010972.4 XP_004011021.4 4 - 5 761 81

View Full Product Details