Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FDXACB1
Entrez Gene ID
101109734
Gene Name
ferredoxin-fold anticodon binding domain containing 1
Gene Aliases
-
Chromosome Location
Chr.15: 24735707 - 24740677 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004016016.4 XP_004016065.2 4 - 5 710 88

View Full Product Details