Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CIDEA
Entrez Gene ID
101108619
Gene Name
cell death inducing DFFA like effector a
Gene Aliases
-
Chromosome Location
Chr.23: 48267310 - 48277937 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004020511.4 XP_004020560.2 2 - 3 239 86
XM_012103593.3 XP_011958983.2 2 - 3 641 86
XM_012103594.3 XP_011958984.2 3 - 4 455 86
XM_012103597.3 XP_011958987.2 2 - 3 671 86

View Full Product Details