Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM3D
Entrez Gene ID
101108169
Gene Name
FAM3 metabolism regulating signaling molecule D
Gene Aliases
-
Chromosome Location
Chr.19: 44673088 - 44723095 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012099781.3 XP_011955171.1 8 - 9 648 68
XM_012099782.3 XP_011955172.1 8 - 9 678 68
XM_012099784.2 XP_011955174.1 8 - 9 648 68

View Full Product Details