Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYNGR3
Entrez Gene ID
101113121
Gene Name
synaptogyrin 3
Gene Aliases
-
Chromosome Location
Chr.24: 1502519 - 1506700 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015104019.2 XP_014959505.2 1 - 2 197 93

View Full Product Details