Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HOXC13
Entrez Gene ID
101108645
Gene Name
homeobox C13
Gene Aliases
-
Chromosome Location
Chr.3: 142107536 - 142115277 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015094610.2 XP_014950096.2 1 - 2 737 95

View Full Product Details