Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC22A20P
Entrez Gene ID
101121727
Gene Name
solute carrier family 22 member 20, pseudogene
Gene Aliases
SLC22A20
Chromosome Location
Chr.21: 45224697 - 45245718 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027959621.1 XP_027815422.1 2 - 3 479 86

View Full Product Details