Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GLIPR1L1
Entrez Gene ID
101121503
Gene Name
GLIPR1 like 1
Gene Aliases
-
Chromosome Location
Chr.3: 118491506 - 118527583 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004006199.4 XP_004006248.3 2 - 3 425 150

View Full Product Details