Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPS19BP1
Entrez Gene ID
101107345
Gene Name
ribosomal protein S19 binding protein 1
Gene Aliases
-
Chromosome Location
Chr.3: 231781785 - 231785437 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004007001.4 XP_004007050.1 2 - 3 286 67

View Full Product Details