Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC101118109
Entrez Gene ID
101118109
Gene Name
protein THEMIS
Gene Aliases
THEMIS
Chromosome Location
Chr.8: 58855296 - 59061495 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004011308.4 XP_004011357.2 4 - 5 1859 111

View Full Product Details