Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRSS27
Entrez Gene ID
101117468
Gene Name
serine protease 27
Gene Aliases
-
Chromosome Location
Chr.24: 2070889 - 2078973 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012103989.2 XP_011959379.2 2 - 3 251 93

View Full Product Details