Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRMT6
Entrez Gene ID
101119859
Gene Name
protein arginine methyltransferase 6
Gene Aliases
-
Chromosome Location
Chr.1: 88917572 - 88919873 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004002251.4 XP_004002300.1 1 - 1 970 68

View Full Product Details