Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBM43
Entrez Gene ID
101119882
Gene Name
RNA binding motif protein 43
Gene Aliases
-
Chromosome Location
Chr.2: 169969574 - 169979543 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004004707.4 XP_004004756.1 2 - 3 375 92

View Full Product Details