Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLA2G12A
Entrez Gene ID
101119561
Gene Name
phospholipase A2 group XIIA
Gene Aliases
-
Chromosome Location
Chr.6: 18790410 - 18803576 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004009623.4 XP_004009672.1 1 - 2 366 63

View Full Product Details