Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTIF3
Entrez Gene ID
101115042
Gene Name
mitochondrial translational initiation factor 3
Gene Aliases
-
Chromosome Location
Chr.10: 34291298 - 34305254 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004012154.4 XP_004012203.3 5 - 6 939 76
XM_015098160.2 XP_014953646.1 5 - 6 973 76
XM_015098161.2 XP_014953647.1 5 - 6 1057 76
XM_015098162.2 XP_014953648.1 5 - 6 911 76
XM_015098163.2 XP_014953649.1 6 - 7 1077 76
XM_015098164.2 XP_014953650.1 6 - 7 1016 76
XM_027973640.1 XP_027829441.1 6 - 7 1112 76
XM_027973641.1 XP_027829442.1 6 - 7 1049 76
XM_027973643.1 XP_027829444.1 6 - 7 988 76

View Full Product Details