Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD61
Entrez Gene ID
101115847
Gene Name
ankyrin repeat domain 61
Gene Aliases
-
Chromosome Location
Chr.24: 38776292 - 38780557 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004021055.4 XP_004021104.3 1 - 2 651 104

View Full Product Details