Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLK7
Entrez Gene ID
101115562
Gene Name
kallikrein related peptidase 7
Gene Aliases
-
Chromosome Location
Chr.14: 60464311 - 60469400 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004023371.4 XP_004023420.3 4 - 5 605 85
XM_027977523.1 XP_027833324.1 3 - 4 894 85

View Full Product Details