Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC101119839
Entrez Gene ID
101119839
Gene Name
signal-regulatory protein beta-2
Gene Aliases
SIRPD
Chromosome Location
Chr.13: 61997615 - 62006173 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027976439.1 XP_027832240.1 3 - 4 778 89

View Full Product Details