Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BTLA
Entrez Gene ID
101120975
Gene Name
B and T lymphocyte associated
Gene Aliases
-
Chromosome Location
Chr.1: 194873074 - 194908008 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027979935.1 XP_027835736.1 3 - 4 587 91

View Full Product Details