Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TPM4
Entrez Gene ID
101117499
Gene Name
tropomyosin 4
Gene Aliases
-
Chromosome Location
Chr.5: 7048114 - 7072856 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004008453.3 XP_004008502.1 2 - 3 287 90
XM_004008454.3 XP_004008503.1 2 - 3 293 90

View Full Product Details