Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHGB
Entrez Gene ID
101115639
Gene Name
chromogranin B
Gene Aliases
-
Chromosome Location
Chr.13: 49973182 - 49990671 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027976824.1 XP_027832625.1 3 - 4 615 103
XM_027976825.1 XP_027832626.1 4 - 5 435 103

View Full Product Details