Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRMT61B
Entrez Gene ID
101102217
Gene Name
tRNA methyltransferase 61B
Gene Aliases
-
Chromosome Location
Chr.3: 37297577 - 37312739 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027966644.1 XP_027822445.1 2 - 3 830 96

View Full Product Details