Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KRT32
Entrez Gene ID
100526788
Gene Name
keratin 32
Gene Aliases
-
Chromosome Location
Chr.11: 21455832 - 21463733 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004012899.4 XP_004012948.2 4 - 5 829 89

View Full Product Details