Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC114108625
Entrez Gene ID
114108625
Gene Name
uncharacterized LOC114108625
Gene Aliases
-
Chromosome Location
Chr.20: 54361311 - 54409781 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027958718.1 XP_027814519.1 1 - 2 996 59

View Full Product Details