Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC105603910
Entrez Gene ID
105603910
Gene Name
myosin light chain kinase family member 4
Gene Aliases
MYLK4
Chromosome Location
Chr.20: 54410617 - 54430240 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027958720.1 XP_027814521.1 3 - 4 492 91

View Full Product Details