Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BMP10
Entrez Gene ID
101109160
Gene Name
bone morphogenetic protein 10
Gene Aliases
-
Chromosome Location
Chr.3: 41277147 - 41284045 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004005819.4 XP_004005868.2 1 - 2 337 65

View Full Product Details