Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HMGN5
Entrez Gene ID
101108552
Gene Name
high mobility group nucleosome binding domain 5
Gene Aliases
-
Chromosome Location
Chr.X: 70314321 - 70322498 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012106988.2 XP_011962378.1 2 - 3 168 72

View Full Product Details