Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM8A1
Entrez Gene ID
101112268
Gene Name
family with sequence similarity 8 member A1
Gene Aliases
-
Chromosome Location
Chr.20: 43147529 - 43158476 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015102838.2 XP_014958324.2 4 - 5 1192 94

View Full Product Details