Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GINS3
Entrez Gene ID
101105096
Gene Name
GINS complex subunit 3
Gene Aliases
-
Chromosome Location
Chr.14: 27801238 - 27807169 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004015038.4 XP_004015087.1 1 - 2 536 89

View Full Product Details