Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NKAPD1
Entrez Gene ID
101111224
Gene Name
NKAP domain containing 1
Gene Aliases
C15H11orf57
Chromosome Location
Chr.15: 24891561 - 24903872 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004016022.4 XP_004016071.1 5 - 6 465 128
XM_027979193.1 XP_027834994.1 5 - 6 974 128

View Full Product Details