Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZKSCAN2
Entrez Gene ID
101108275
Gene Name
zinc finger with KRAB and SCAN domains 2
Gene Aliases
-
Chromosome Location
Chr.24: 23481983 - 23498785 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027961435.1 XP_027817236.1 1 - 2 768 107

View Full Product Details