Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PM20D2
Entrez Gene ID
101111105
Gene Name
peptidase M20 domain containing 2
Gene Aliases
-
Chromosome Location
Chr.8: 53890092 - 53903348 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015097387.2 XP_014952873.2 5 - 6 1095 94

View Full Product Details