Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PASD1
Entrez Gene ID
101106666
Gene Name
PAS domain containing repressor 1
Gene Aliases
-
Chromosome Location
Chr.X: 91073626 - 91143752 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027963456.1 XP_027819257.1 1 - 2 76 107

View Full Product Details