Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CA9
Entrez Gene ID
101115682
Gene Name
carbonic anhydrase 9
Gene Aliases
-
Chromosome Location
Chr.2: 55576428 - 55584905 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004004279.4 XP_004004328.1 3 - 4 2335 92
XM_012121475.3 XP_011976865.1 3 - 4 2335 92

View Full Product Details