Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCAS4
Entrez Gene ID
101118977
Gene Name
breast carcinoma amplified sequence 4
Gene Aliases
-
Chromosome Location
Chr.13: 82149720 - 82209649 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012189344.3 XP_012044734.2 1 - 2 167 88

View Full Product Details