Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMIGD2
Entrez Gene ID
101115367
Gene Name
transmembrane and immunoglobulin domain containing 2
Gene Aliases
-
Chromosome Location
Chr.5: 18253823 - 18261537 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004009357.4 XP_004009406.2 4 - 5 945 78

View Full Product Details