Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NAGS
Entrez Gene ID
101112658
Gene Name
N-acetylglutamate synthase
Gene Aliases
-
Chromosome Location
Chr.11: 19191672 - 19197350 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027974449.1 XP_027830250.1 3 - 4 945 82

View Full Product Details