Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DFFA
Entrez Gene ID
101114715
Gene Name
DNA fragmentation factor subunit alpha
Gene Aliases
-
Chromosome Location
Chr.12: 46508863 - 46523102 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004013730.4 XP_004013779.1 2 - 3 385 101
XM_012187337.3 XP_012042727.1 2 - 3 385 101

View Full Product Details