Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AZGP1
Entrez Gene ID
101123437
Gene Name
alpha-2-glycoprotein 1, zinc-binding
Gene Aliases
-
Chromosome Location
Chr.24: 37128167 - 37132700 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012104859.3 XP_011960249.3 2 - 3 453 100

View Full Product Details