Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATCAY
Entrez Gene ID
101116652
Gene Name
ATCAY kinesin light chain interacting caytaxin
Gene Aliases
-
Chromosome Location
Chr.5: 18547719 - 18587835 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027969868.1 XP_027825669.1 3 - 4 549 54

View Full Product Details