Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NAPSA
Entrez Gene ID
101116503
Gene Name
napsin A aspartic peptidase
Gene Aliases
-
Chromosome Location
Chr.14: 59949696 - 59956856 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012190732.3 XP_012046122.2 2 - 3 494 80

View Full Product Details